Merck
  • Methylation damage to RNA induced in vivo in Escherichia coli is repaired by endogenous AlkB as part of the adaptive response.
Methylation damage to RNA induced in vivo in Escherichia coli is repaired by endogenous AlkB as part of the adaptive response.
DNA repair (2013-01-02)
Cathrine Broberg Vågbø, Eva K Svaasand, Per A Aas, Hans E Krokan
ABSTRACT
Cytotoxic 1-methyladenine (1-meA) and 3-methylcytosine (3-meC) lesions induced in DNA and RNA in vitro and in pre-damaged DNA and RNA bacteriophages in vivo are repaired by the Escherichia coli (E. coli) protein AlkB and a human homolog, ALKBH3. However, it is not known whether endogenous RNA is repaired in vivo by repair proteins present at physiological concentrations. The concept of RNA repair as a biologically relevant process has therefore remained elusive. Here, we demonstrate AlkB-mediated repair of endogenous RNA in vivo by measuring differences in lesion-accumulation in two independent AlkB-proficient and deficient E. coli strains during exposure to methyl methanesulfonate (MMS). Repair was observed both in AlkB-overproducing strains and in the wild-type strains after AlkB induction. RNA repair appeared to be highest in RNA species below 200 nucleotides in size, mainly comprising tRNAs. Strikingly, at least 10-fold more lesions were repaired in RNA than in DNA. This may be a consequence of some 30-fold higher levels of aberrant methylation in RNA than in DNA after exposure to MMS. A high primary kinetic isotope effect (>10) was measured using a deuterated methylated RNA substrate, D3-1me(rA), demonstrating that it is the catalytic step, and not the search step that is rate-limiting. Our results demonstrate that RNA repair by AlkB takes place in endogenous RNA as part of an adaptive response in wild-type E. coli cells.
Sigma-Aldrich
Methyl methanesulfonate, 99%