- Rhodium complex immobilized on graphene oxide as an efficient and recyclable catalyst for hydrogenation of cyclohexene.
Rhodium complex immobilized on graphene oxide as an efficient and recyclable catalyst for hydrogenation of cyclohexene.
Nanoscale (2012-12-15)
Qingshan Zhao, Dafa Chen, Yang Li, Guoliang Zhang, Fengbao Zhang, Xiaobin Fan
PMID23238302
ABSTRACT
Rhodium complexes can be homogeneously immobilized on functionalized graphene oxide through coordination interaction. The obtained catalyst can be readily recycled and shows enhanced activity in the catalytic hydrogenation of cyclohexene.