Merck
  • Selective, sensitive and reversible "turn-on" fluorescent cyanide probes based on 2,2'-dipyridylaminoanthracene-Cu2+ ensembles.
Selective, sensitive and reversible "turn-on" fluorescent cyanide probes based on 2,2'-dipyridylaminoanthracene-Cu2+ ensembles.
Chemical communications (Cambridge, England) (2012-10-24)
Yongshu Xie, Yubin Ding, Xin Li, Cheng Wang, Jonathan P Hill, Katsuhiko Ariga, Weibing Zhang, Weihong Zhu
ABSTRACT
2,2'-Dipyridylamine and anthracene units were linked to afford highly emissive compounds whose Cu(2+) ensembles were developed as effective fluorescence turn-on CN(-) probes.
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, reagent grade, 97%