Merck
  • Reaction kinetics of selected micropollutants in ozonation and advanced oxidation processes.
Reaction kinetics of selected micropollutants in ozonation and advanced oxidation processes.
Water research (2012-10-20)
Xiaohui Jin, Sigrid Peldszus, Peter M Huck
ABSTRACT
Second-order reaction rate constants of micropollutants with ozone (k(O3)) and hydroxyl radicals (k(OH)) are essential for evaluating their removal efficiencies from water during ozonation and advanced oxidation processes. Kinetic data are unavailable for many of the emerging micropollutants. Twenty-four micropollutants with very diverse structures and applications including endocrine disrupting compounds, pharmaceuticals, and personal care products were selected, and their k(O3) and k(OH) values were determined using bench-scale reactors (at pH 7 and T = 20 °C). Reactions with molecular ozone are highly selective as indicated by their k(O3) values ranging from 10(-2)-10(7) M(-1) s(-1). The general trend of ozone reactivity can be explained by micropollutant structures in conjunction with the electrophilic nature of ozone reactions. All of the studied compounds are highly reactive with hydroxyl radicals as shown by their high k(OH) values (10(8)-10(10) M(-1) s(-1)) even though they are structurally very diverse. For compounds with a low reactivity toward ozone, hydroxyl radical based treatment such as O(3)/H(2)O(2) or UV/H(2)O(2) is a viable alternative. This study contributed to filling the data gap pertaining kinetic data of organic micropollutants while confirming results reported in the literature where available.
Product Description
Supelco
Anisole, analytical standard
Sigma-Aldrich
Anisole, ReagentPlus®, 99%
Supelco
Anisole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anisole, anhydrous, 99.7%
Sigma-Aldrich
Anisole, ≥99%, FCC, FG