Merck
  • Acute and subacute toxicity of novel biogenic selenium nanoparticles in mice.
Acute and subacute toxicity of novel biogenic selenium nanoparticles in mice.
Pharmaceutical biology (2012-10-06)
Mojtaba Shakibaie, Ahmad Reza Shahverdi, Mohammad Ali Faramarzi, Gholam Reza Hassanzadeh, Hamid Reza Rahimi, Omid Sabzevari
ABSTRACT
In the present investigation, acute and subacute toxicity of the biogenic Se nanoparticles (Se NPs) has been reported. To characterize the Se NPs produced by a bacterium species and to evaluate their toxicity and impact on clinical chemistry and hematological parameters of NMRI mice. The Se NPs were prepared by Bacillus sp. MSh-1 in a culture medium containing SeO(2) (1.26 mM) and their physiochemical properties investigated using TEM, XRD and FT-IR. The LD(50) of Se NPs and SeO(2) were determined and the subacute toxicity evaluated by orally administration of 0, 2.5, 5, 10 and 20 mg kg(-1) of Se NPs to male mice for 14 consecutive days. Parameters of blood cells, AST, ALT, ALP, creatinine, BUN, cholesterol, bilirubin, triglyceride and CPK were experimentally measured. The XRD and TEM analyses showed that the spherical NPs were amorphous, in the size range of 80-220 nm. The toxicological evaluation showed that the LD(50) values of SeO(2) and Se NPs were 7.3 and 198.1 mg kg(-1), respectively. No biochemical changes were observed from the administration of 2.5, 5 and 10 mg kg(-1) of Se NPs, but a dose of 20 mg kg(-1) was accompanied with signs of toxicity including lower body weight and changes in clinical chemistry and hematological parameters. The biogenic Se NPs were less toxic than synthetic Se NPs and much less (26-fold) toxic than the SeO(2), which demonstrates the important role of Bacillus sp. MSh-1 in conversion of a highly toxic Se compound to the less toxic Se NPs.
Sigma-Aldrich
Selenium dioxide, ReagentPlus®, powder, 99.8% trace metals basis
Sigma-Aldrich
Selenium dioxide, reagent grade, powder, 98%
Sigma-Aldrich
Selenium dioxide, ≥99.9% trace metals basis
Sigma-Aldrich
Selenium dioxide, 99.999% trace metals basis