Merck
  • Palladium-catalyzed nitromethylation of aryl halides: an orthogonal formylation equivalent.
Palladium-catalyzed nitromethylation of aryl halides: an orthogonal formylation equivalent.
Organic letters (2012-07-31)
Ryan R Walvoord, Simon Berritt, Marisa C Kozlowski
ABSTRACT
An efficient cross-coupling reaction of aryl halides and nitromethane was developed with the use of parallel microscale experimentation. The arylnitromethane products are precursors for numerous useful synthetic products. An efficient method for their direct conversion to the corresponding oximes and aldehydes in a one-pot operation has been discovered. The process exploits inexpensive nitromethane as a carbonyl equivalent, providing a mild and convenient formylation method that is compatible with many functional groups.
Sigma-Aldrich
Nitromethane, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Nitromethane, suitable for HPLC, ≥96%
Sigma-Aldrich
Nitromethane, puriss., absolute, over molecular sieve, ≥98.5% (GC)
Sigma-Aldrich
Nitromethane, ACS reagent, ≥95%