Merck
  • Fluorimetric determination of thiabendazole residues in mushrooms using sequential injection analysis.
Fluorimetric determination of thiabendazole residues in mushrooms using sequential injection analysis.
Talanta (2012-07-24)
E J Llorent-Martínez, M L Fernández-de Córdova, A Ruiz-Medina, P Ortega-Barrales
ABSTRACT
Thiabendazole is a benzimidazole fungicide of general use that is specifically used to control mushroom diseases, mainly cobweb diseases, which is caused by members of the genus Cladobotryum. Although this compound is legislated and its maximum residue limit established at 60mgkg(-1) by Codex Alimentarius, there is almost a complete absence of analytical methods available for its determination in mushrooms. Here, we propose an automated method, using Sequential Injection Analysis with fluorescence detection (λ(exc)/λ(em)=305/345nm) for the determination of thiabendazole in mushrooms. We have developed a flow-through optosensor using C(18) silica gel as solid support placed in the flow-cell where the determination is performed. This method presents a detection limit of 0.5mgkg(-1), and recovery experiments have been carried out in different kinds of mushrooms at levels below the legislated maximum residue limit, demonstrating that the proposed analytical method fulfils the requirements for its applications in quality control of mushrooms.
Product Description
Supelco
Thiabendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thiabendazole, suitable for plant cell culture
Sigma-Aldrich
Thiabendazole, ≥99%, powder
Supelco
Thiabendazole, PESTANAL®, analytical standard
Tiabendazole, European Pharmacopoeia (EP) Reference Standard