Merck
  • Prophylactic zinc supplementation reduces bacterial load and improves survival in a murine model of sepsis.
Prophylactic zinc supplementation reduces bacterial load and improves survival in a murine model of sepsis.
Pediatric critical care medicine : a journal of the Society of Critical Care Medicine and the World Federation of Pediatric Intensive and Critical Care Societies (2012-07-05)
Jeffrey E Nowak, Kelli Harmon, Charles C Caldwell, Hector R Wong
ABSTRACT
We previously demonstrated that altered zinc homeostasis is an important feature of pediatric sepsis, thus raising the possibility of zinc supplementation as a therapeutic strategy in sepsis. Herein, we tested the hypothesis that prophylactic zinc supplementation would be beneficial in a murine model of peritoneal sepsis. Murine model of sepsis (intraperitoneal fecal-slurry injection). Basic science research laboratory. C57BL/6 male mice. Intraperitoneal fecal-slurry injection, with or without zinc supplementation (10 mg/kg of intraperitoneal zinc gluconate for 3 days prior to intraperitoneal fecal-slurry injection). Survival over 3 days following intraperitoneal fecal-slurry injection, markers of inflammation, bacterial load studies, and immunophenotyping studies. Zinc-supplemented mice demonstrated a significant survival advantage compared to control (nonsupplemented) mice. Zinc-supplemented mice also demonstrated moderate reductions of inflammation and immune activation. The survival advantage primarily correlated with reduced in vivo bacterial load in zinc-supplemented mice, compared to controls. In addition, peritoneal macrophages harvested from zinc-supplemented mice demonstrated a significantly enhanced phagocytosis capacity for Escherichia coli and Staphylococcus aureus, compared to peritoneal macrophages harvested from control mice. Prophylactic zinc supplementation reduces bacterial load and is beneficial in a murine model of peritoneal sepsis.
Product Description
Supelco
D-Gluconic acid sodium salt, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Sigma-Aldrich
D-Gluconic acid sodium salt, ≥99% (HPLC)
Sigma-Aldrich
Sodium gluconate, meets USP testing specifications
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(II) D-gluconate dihydrate, 98%
Sigma-Aldrich
Magnesium D-gluconate hydrate, ≥98% (HPLC)
Sigma-Aldrich
Magnesium D-gluconate hydrate, 98.0-102% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)