Merck
  • Tunable synthesis of metal-graphene complex nanostructures and their catalytic ability for solvent-free cyclohexene oxidation in air.
Tunable synthesis of metal-graphene complex nanostructures and their catalytic ability for solvent-free cyclohexene oxidation in air.
Nanoscale (2012-06-15)
Hui Huang, Hengchao Zhang, Zheng Ma, Yang Liu, Hai Ming, Haitao Li, Zhenhui Kang
ABSTRACT
A one-step method was developed for the controllable construction of metal-graphene core-shell structures, hollow graphene nanospheres, and a high density of metal nanoparticles supported on graphene. The metal-graphene core-shell nanostructures as nanocatalysts show excellent catalytic ability for the selective oxidation of cyclohexene.
Product Description
Supelco
Cyclohexene, analytical standard, ≥99.5% (GC)
Sigma-Aldrich
Cyclohexene, contains 100 ppm BHT as inhibitor, ≥99.0%
Sigma-Aldrich
Cyclohexene, inhibitor-free, ReagentPlus®, 99%