Merck
Decarboxylative etherification of aromatic carboxylic acids.
Journal of the American Chemical Society (2012-06-12)
Sukalyan Bhadra, Wojciech I Dzik, Lukas J Goossen
ABSTRACT
Decarboxylative Chan-Evans-Lam-type couplings are presented as a new strategy for the regiospecific construction of diaryl and alkyl aryl ethers starting from easily available aromatic carboxylic acids. They allow converting various aromatic carboxylate salts into the corresponding aryl ethers by reaction with alkyl orthosilicates or aryl borates, under aerobic conditions in the presence of silver carbonate as the decarboxylation catalyst and copper acetate as the cross-coupling catalyst.
Sigma-Aldrich
Silver carbonate on Celite®, extent of labeling: ~50 wt. % loading
Sigma-Aldrich
Silver carbonate, purum p.a., ≥99.0% (AT)
Sigma-Aldrich
Silver carbonate, 99%