Merck
  • [Biosynthesis of geosmin and 2-methylisoborneol in the prokaryotes--a review].
[Biosynthesis of geosmin and 2-methylisoborneol in the prokaryotes--a review].
Wei sheng wu xue bao = Acta microbiologica Sinica (2012-05-17)
Ting Zhang, Deliang Li, Jie Li
ABSTRACT
The occurrence of off-flavor problems caused by prokaryotes cyanobacteria and actinomycetes is a worldwide water and food quality issue. Based on literatures on two earthy-muddy-smelling metabolites, (i. e., geosmin and 2-methylisoborneol (2-MIB)), we reviewed their chemical characteristics, biosynthetic pathways, genes and enzymes that are involved in biosynthesis. Then we discussed current research questions related to off-flavor and future directions. Finally, we addressed the significance of probable key regulatory mechanism for the production and release of geosmin and 2-MIB, which could provide more scientific strategies to better control off-flavors outbreaks in drinking and aquaculture water.
Product Description
Supelco
2-Methylisoborneol solution, certified reference material, TraceCERT®, 100 μg/mL in methanol, ampule of 1 mL
Sigma-Aldrich
2-Methylisoborneol solution, ~10 mg/mL in methanol, ≥98% (GC)