Merck
  • Silicon determination in human ventricular whole blood: a possible marker of drowning.
Silicon determination in human ventricular whole blood: a possible marker of drowning.
Analytical biochemistry (2012-05-01)
Federica Maraschi, Michela Sturini, Andrea Speltini, Francesco Orio, Antonella Profumo, Giovanni Pierucci
ABSTRACT
This article presents the first results demonstrating that total silicon trace concentration in human ventricular whole blood may be used as a further marker in the diagnosis of drowning. The difference in silicon content between the left and right ventricles was significantly higher for drowning cases than that from individuals who had not drowned. These findings were in full agreement with autoptic responses, supporting silicon as a marker of freshwater drowning. The procedure entails an alkaline microwave-assisted digestion using tetramethylammonium hydroxide (TMAH) in the presence of H(2)O(2) followed by dynamic reaction cell inductively coupled plasma mass spectrometry (DRC-ICP-MS) detection, whose accuracy was obtained for Seronorm whole blood reference material. Satisfactory recoveries (91-98%) were gained on whole ventricular blood, with a silicon content lower than the method detection limit (MDL), spiked at 5 to 7μgg(-1) with materials consistent with drowning media constituents, that is, freshwater plankton (CRM [certified reference material] 414), silicon dioxide, diatomaceous earth powder, and a silicon standard solution. Good within-lab reproducibility (4-10%) and sensitivity (MDL=0.46μgg(-1)) were achieved as well. The procedure was applied to blood samples from 18 different real cases of death.
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Celite® 512 medium, filter aid, calcined
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%