Merck
  • Controllable synthesis of MnS nanocrystals from a single-source precursor.
Controllable synthesis of MnS nanocrystals from a single-source precursor.
Journal of colloid and interface science (2012-04-21)
Long Peng, Shuling Shen, Yejun Zhang, Huarui Xu, Qiangbin Wang
ABSTRACT
A facile single-source precursor method has been applied for the selective synthesis of MnS nanocrystals (NCs) with well-defined shapes and crystal structures such as hexapod, octahedral, hexagonal shaped α-MnS NCs, and pencil-shaped γ-MnS NCs. The effects of the composition of precursor, reaction temperature, and the heating rate on the morphologies, and crystal structures of MnS NCs were systematically studied for the first time.
Sigma-Aldrich
Manganese sulfate monohydrate, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, BioReagent, suitable for plant cell culture, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, puriss., meets analytical specification of Ph. Eur, BP, USP, FCC, 99-100.5% (calc. for dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese(II) sulfate monohydrate, ACS reagent, ≥98%
Pricing and availability is not currently available.