Merck
  • Comparison of phenolic acids profile and antioxidant potential of six varieties of spelt (Triticum spelta L.).
Comparison of phenolic acids profile and antioxidant potential of six varieties of spelt (Triticum spelta L.).
Journal of agricultural and food chemistry (2012-04-17)
Urszula Gawlik-Dziki, Michał Świeca, Dariusz Dziki
ABSTRACT
Phenolic acids profile and antioxidant activity of six diverse varieties of spelt are reported. Antioxidant activity was assessed using eight methods based on different mechanism of action. Phenolic acids composition of spelt differed significantly between varieties and ranged from 506.6 to 1257.4 μg/g DW. Ferulic and sinapinic acids were the predominant phenolic acids found in spelt. Total ferulic acid content ranged from 144.2 to 691.5 μg/g DW. All analyzed spelt varieties possessed high antioxidant potential. In spite of the fact that bound phenolic acids possessed higher antioxidant activities, analysis of antioxidant potential and their relationship with phenolic acid content showed that free phenolics were more effective. Eight antioxidant methods were integrated to obtain a total antioxidant capacity index that may be used for comparison of total antioxidant capacity of spelt varieties. Total antioxidant potential of spelt cultivars were ordered as follows: Ceralio > Spelt INZ ≈ Ostro > Oberkulmer Rotkorn > Schwabenspelz > Schwabenkorn.
Product Description
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure