Merck
  • Major complications, mortality, and resource utilization after open abdominal surgery: 0.9% saline compared to Plasma-Lyte.
Major complications, mortality, and resource utilization after open abdominal surgery: 0.9% saline compared to Plasma-Lyte.
Annals of surgery (2012-04-04)
Andrew D Shaw, Sean M Bagshaw, Stuart L Goldstein, Lynette A Scherer, Michael Duan, Carol R Schermer, John A Kellum
ABSTRACT
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace volume losses after major surgery. An observational study using the Premier Perspective Comparative Database was performed to evaluate adult patients undergoing major open abdominal surgery who received either 0.9% saline (30,994 patients) or a balanced crystalloid solution (926 patients) on the day of surgery. The primary outcome was major morbidity and secondary outcomes included minor complications and acidosis-related interventions. Outcomes were evaluated using multivariable logistic regression and propensity scoring models. For the entire cohort, the in-hospital mortality was 5.6% in the saline group and 2.9% in the balanced group (P < 0.001). One or more major complications occurred in 33.7% of the saline group and 23% of the balanced group (P < 0.001). In the 3:1 propensity-matched sample, treatment with balanced fluid was associated with fewer complications (odds ratio 0.79; 95% confidence interval 0.66-0.97). Postoperative infection (P = 0.006), renal failure requiring dialysis (P < 0.001), blood transfusion (P < 0.001), electrolyte disturbance (P = 0.046), acidosis investigation (P < 0.001), and intervention (P = 0.02) were all more frequent in patients receiving 0.9% saline. Among hospitals in the Premier Perspective Database, the use of a calcium-free balanced crystalloid for replacement of fluid losses on the day of major surgery was associated with less postoperative morbidity than 0.9% saline.
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)