Merck
  • Applications of Hadamard transform-gas chromatography/mass spectrometry for the detection of hexamethyldisiloxane in a wafer cleanroom.
Applications of Hadamard transform-gas chromatography/mass spectrometry for the detection of hexamethyldisiloxane in a wafer cleanroom.
Journal of chromatography. A (2011-12-24)
Yuan-Kai Cheng, Cheng-Huang Lin, Samuel Kuo, Jonathan Yang, Szu-Yuan Hsiung, Jia-Lin Wang
ABSTRACT
The Hadamard transform-gas chromatography/mass spectrometry (HT-GC/MS) technique was successfully employed for the detection of hexamethyldisiloxane (HMDSO, C(6)H(18)OSi(2)) at the sub-nL/L level in a semiconductor wafer cleanroom. Indoor air samples were collected from the room, according to EPA Method TO-17 using a Tedlar bag where the air samples were allowed to pass through an absorption tube for 24 h. The condensed components were then heated and simultaneously injected into a GC column through a Hadamard-injector, which was operated in accordance with the Hadamard codes. Compared to the single injection used in most GC/MS systems, the signal-to-noise (S/N) ratios were substantially improved after the inverse Hadamard transformation of the encoded chromatogram. Under optimized conditions, when cyclic S-matrix orders of 255, 1023 and 2047 were used, the S/N ratios of the HMDSO signals were substantially improved by 7.4-, 15.1- and 20.1-fold, respectively. These improvements are in good agreement with theoretically calculated values (8.0-, 16.0- and 22.6-fold, respectively). We found that when the HT-GC/MS technique was applied, HMDSO could be detected at the 0.1 nL/L level.
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Hexamethyldisiloxane, ≥98%
Sigma-Aldrich
Hexamethyldisiloxane, NMR grade, ≥99.5%
Sigma-Aldrich
Hexamethyldisiloxane, puriss., ≥98.5% (GC)