Merck
  • Utilization of chicken feather hydrolysate as a novel fermentation substrate for production of exopolysaccharide and mycelial biomass from edible mushroom Morchella esculenta.
Utilization of chicken feather hydrolysate as a novel fermentation substrate for production of exopolysaccharide and mycelial biomass from edible mushroom Morchella esculenta.
International journal of food sciences and nutrition (2011-12-06)
Mesut Taskin, Behzat Ozkan, Okkes Atici, Mehmet Nuri Aydogan
ABSTRACT
This study was performed to investigate the usability of chicken feather hydrolysate (Chicken feather peptone (CFP)) as substrate for mycelial biomass and extracellular polysaccharides (EPS) production from edible mushroom Morchella esculenta. The ability of CFP to support biomass and EPS production in edible mushroom M. esculenta was compared to those of two commercial peptones (Tryptone peptone (TP) and Fish peptone (FP)). The maximum biomass (16.3 g/l) and EPS (4.8 g/l) concentrations were achieved with TP. Second, high biomass (15.9 g/l) and EPS (4.6 g/l) concentrations were obtained with CFP. Also, biomass and EPS concentrations in CFP medium were statistically near to those in the TP medium. CFP and TP resulted in not only uniform pellets with smaller size (5 mm) but also faster mycelial growth compared to FP. This study showed for the first time that CFP could be effectively used as a novel EPS production substrate.
Product Description
Millipore
Peptone from milk solids, Refined hydrolysate
Millipore
Proteose Peptone, Enzymatic hydrolysate
Millipore
Peptone from casein and other animal proteins, suitable for microbiology
Millipore
Peptone from animal tissue, from meat
Millipore
Peptone Primatone® RL, suitable for microbiology
Sigma-Aldrich
Peptone from animal tissue, from meat, BioReagent, suitable for cell culture, suitable for plant cell culture