Merck
  • Rapid determination of terephthalic acid in the hydrothermal decomposition product of poly(ethylene terephthalate) by thermochemolysis-gas chromatography in the presence of tetramethylammonium acetate.
Rapid determination of terephthalic acid in the hydrothermal decomposition product of poly(ethylene terephthalate) by thermochemolysis-gas chromatography in the presence of tetramethylammonium acetate.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2011-10-12)
Yasuyuki Ishida, Kei Ohsugi, Kohei Taniguchi, Hajime Ohtani
ABSTRACT
Thermochemolysis-gas chromatography in the presence of tetramethylammonium acetate was applied to the direct determination of terephthalic acid (TPA) contained in solid decomposition products obtained from the hydrothermal recycling process of poly(ethylene terephthalate) (PET). On the chromatograms of the hydrothermal decomposition products of PET, a sharp peak of the TPA component was clearly observed as its corresponding dimethyl ester formed through the thermochemolysis reaction. Based on the peak intensities, the contents of TPA in the decomposition products were determined precisely and rapidly without using any cumbersome sample pretreatments.
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%