Merck
  • Accelerating palladium-catalyzed C-F bond formation: use of a microflow packed-bed reactor.
Accelerating palladium-catalyzed C-F bond formation: use of a microflow packed-bed reactor.
Angewandte Chemie (International ed. in English) (2011-08-13)
Timothy Noël, Thomas J Maimone, Stephen L Buchwald
ABSTRACT
[Image: see text] A flow process for Pd-catalyzed carbon fluorine bond formation is described. A microreactor using a packed-bed design allows for easy handling of large quantities of insoluble CsF with precise control over reaction times, efficient mixing, and the ability to safely handle elevated temperatures and pressures. A variety of aryl triflates, including heteroaryl ones, were converted to their corresponding aryl fluoride in short reaction times that would be difficult to achieve in a typical batch process.
Sigma-Aldrich
Cesium Fluoride ChemBeads, NSC 84270
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%