Merck
  • A study of parameter setting and characterization of visible-light driven nitrogen-modified commercial TiO2 photocatalysts.
A study of parameter setting and characterization of visible-light driven nitrogen-modified commercial TiO2 photocatalysts.
Journal of hazardous materials (2011-05-11)
Yu-Lin Kuo, Te-Li Su, Fu-Chen Kung, Tsai-Jung Wu
ABSTRACT
An optimal condition applied to the Taguchi method with an L(9) orthogonal array for preparing a visible-light driven nitrogen-modified TiO(2) (N-TiO(2)) photocatalyst by a simple hydrolysis method has been examined for material characteristics and a photodecolorization test of methyl blue (MB) under various visible light source (fluorescent and blue LED lamps) irradiations. Results of the material characterization showed that the absorption of prepared N-TiO(2) powder exhibited a significant extension into visible light regimes with an optical bandgap (Eg) of around 2.96 eV, which subsequently improved the visible-light photocatalytic activity of N-TiO(2) samples. The superior photocatalytic properties, the pseudo first-order reaction rate constants (k) and photodecolorization efficiency (η%) of a N-TiO(2) photocatalyst during the photodecolorization test of methyl blue (MB) under two different visible light irradiations were very evident compared to those for pure TiO(2). For photodecolorization of practical dyeing from the waste water from the dyeing and finishing industry, a higher photodecolorization efficiency of N-TiO(2) powder toward Direct blue-86 (DB-86) (Direct Fast Turquoise Blue GL) dye was also achieved.
Sigma-Aldrich
Methyl Blue, certified by the Biological Stain Commission
Sigma-Aldrich
Methyl Blue, powder or crystals
Sigma-Aldrich
Methyl Blue, for microscopy (Bot., Hist.), indicator (pH 9.4-14.0)