Merck
  • Infrared thermography and odour composition of the Amorphophallus gigas (Araceae) inflorescence: the cooling effect of the odorous liquid.
Infrared thermography and odour composition of the Amorphophallus gigas (Araceae) inflorescence: the cooling effect of the odorous liquid.
Plant biology (Stuttgart, Germany) (2011-04-15)
S Kakishima, Y Terajima, J Murata, H Tsukaya
ABSTRACT
During the second blooming of a cultivated Amorphophallus gigas Teijsm and Binnend in the Botanical Gardens of the University of Tokyo, the surface temperature of the inflorescence was measured using an infrared camera. Contrary to studies of other species in the genus Amorphophallus, the surface of the inflorescence showed only very faint thermogenesis and had a lower temperature than that of the background. This cooling effect appeared to be due to a loss of heat through evaporation, which was caused by the secretion of a very large amount of odorous liquid. Chemical analysis revealed that the major components of this liquid were acetic acid, propionic acid, butyric acid and valeric acids. The composition of the odorous liquid was slightly different between the spathe surface and the sterile appendix. The major component(s) of the odorous material from the spathe was butyric acid, and from the sterile appendix was valeric acids. These components would play dual roles of adding the characteristic smell to the inflorescence and cooling the inflorescence.
Sigma-Aldrich
Valeric acid, ≥99%, FCC, FG
Supelco
Valeric acid, analytical standard
Sigma-Aldrich
Valeric acid, ≥99%