Merck
Total synthesis of (-)-indolactam V.
Organic & biomolecular chemistry (2011-02-23)
Zhengren Xu, Fengying Zhang, Lihe Zhang, Yanxing Jia
ABSTRACT
The total synthesis of protein kinase C activator (-)-indolactam V (IL-V) has been successfully completed with two separate approaches: From known 4-nitrotryptophan derivative 3 in 8 steps (49% overall yield) and from L-glutamic acid in 12 steps (18% overall yield), where 4-nitrotryptophanol derivative 4 served as a key intermediate. Derivatives 3 and 4, both incorporating indole 4-substitution and the C-9 stereocenter in IL-V, were synthesized via the Pd-catalyzed indole synthesis from 3-nitro-2-iodoaniline 5 with aldehydes 6 and 7, respectively. Aldehyde 7 was, meanwhile, synthesized from l-glutamic acid in 5 steps (68% yield). Lactamization of the 9-membered ring was achieved using HATU in THF in good yield.
Sigma-Aldrich
(−)-Indolactam V, ≥96% (HPLC)