Merck
  • Cyclometalated red iridium(III) complexes containing carbazolyl-acetylacetonate ligands: efficiency enhancement in polymer LED devices.
Cyclometalated red iridium(III) complexes containing carbazolyl-acetylacetonate ligands: efficiency enhancement in polymer LED devices.
Dalton transactions (Cambridge, England : 2003) (2010-08-19)
Nan Tian, Yaroslav V Aulin, Daniel Lenkeit, Simon Pelz, Oleksandr V Mikhnenko, Paul W M Blom, Maria Antonietta Loi, Elisabeth Holder
ABSTRACT
The design, synthesis, photophysical and significantly improved electrooptical properties of a series of red emitting cyclometalated iridium(iii) complexes containing carbazolyl-acetylacetonate ligands are described.
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis
Sigma-Aldrich
Manganese(II) acetylacetonate
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%