Merck
  • Caesium fluoride-induced changes in the c.d. spectra of synthetic DNA fragments.
Caesium fluoride-induced changes in the c.d. spectra of synthetic DNA fragments.
International journal of biological macromolecules (1991-02-01)
J Kypr, M Vorlícková, G Zon, R Weiss, L Arnold, J Smrt
ABSTRACT
Ten DNA fragments containing self-complementary alternating sequences of adenine and thymine differing in length and the starting nucleotide were studied by c.d. spectroscopy. It was found that d(TATATATA) but not d(ATATATAT), d(TATATA), d(CTATATAG) or (dT-dA)20 isomerized into the unusual X-DNA double helix at molar concentrations of CsF in solution. But in contrast to poly(dA-dT), the octamer (dT-dA)4, isomerized very slowly, at relatively low CsF concentrations and the isomerization was strongly dependent on the octamer concentration. A model is proposed to account for the observed properties of the B-to-X isomerization on the oligomer level.
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%