Merck
  • Enantioselective Henry addition of methyl 4-nitrobutyrate to aldehydes. Chiral building blocks for 2-pyrrolidinones and other derivatives.
Enantioselective Henry addition of methyl 4-nitrobutyrate to aldehydes. Chiral building blocks for 2-pyrrolidinones and other derivatives.
Organic letters (2010-06-12)
Gonzalo Blay, Víctor Hernández-Olmos, José R Pedro
ABSTRACT
A catalytic highly enantioselective Henry addition of methyl 4-nitrobutyrate to aldehydes using a Cu(II)-amino pyridine complex as catalyst is described. The products resulting from this reaction constitute a new, highly versatile family of chiral building blocks as a result of the presence of three different functional groups on the molecule. These products have been transformed into nonracemic chiral gamma-lactams, 5-hydroxy-5-substituted levulinic acid derivatives, and delta-lactones.
Sigma-Aldrich
2-Pyrrolidinone, purum, ≥98.0% (GC)
Sigma-Aldrich
2-Pyrrolidinone, 99%
Sigma-Aldrich
Methyl 4-nitrobutyrate, 97%
Sigma-Aldrich
2-Pyrrolidinone, ≥99%