Merck
  • Nucleophilic heterocyclic carbene as a novel catalyst for cyclopropanation of cyano acrylates.
Nucleophilic heterocyclic carbene as a novel catalyst for cyclopropanation of cyano acrylates.
Organic & biomolecular chemistry (2010-02-06)
Anabha E Raveendran, Rony Rajan Paul, Eringathodi Suresh, Vijay Nair
ABSTRACT
Nucleophilic heterocyclic carbenes (NHCs) have been used for the first time as catalysts in the cyclopropanation of ethyl cyanocinnamates with phenacyl bromide by Michael-initiated ring-closure (MIRC).
Product Description
Supelco
2-Bromoacetophenone, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
2-Bromoacetophenone, 98%