Merck
  • Beta-lactam antibiotic reduces morphine analgesic tolerance in rats through GLT-1 transporter activation.
Beta-lactam antibiotic reduces morphine analgesic tolerance in rats through GLT-1 transporter activation.
Drug and alcohol dependence (2009-12-17)
Scott M Rawls, Michael Zielinski, Hiren Patel, Steven Sacavage, David A Baron, Digvesh Patel
ABSTRACT
Glutamate transporter subtype 1 (GLT-1) activation is a promising - and understudied - approach for managing aspects of morphine tolerance caused by increased glutamatergic transmission. Identification of beta-lactam antibiotics as pharmaceuticals which activate GLT-1 transporters prompted us to hypothesize that repeated beta-lactam antibiotic (ceftriaxone) administration blocks development of tolerance to morphine antinociception through GLT-1 activation. Here, we injected rats with morphine (10mg/kg, s.c.) twice daily for 7 days to induce tolerance and used the hot-plate assay to determine antinociception on days 1, 4 and 7 of repeated morphine administration. Ceftriaxone and a selective GLT-1 transporter inhibitor dihydrokainate (DHK) were co-administered with morphine to determine if GLT-1 activation mediated the ceftriaxone effect. Tolerance was present on days 4 and 7 of repeated morphine administration. Ceftriaxone (50, 100 or 200mg/kg, i.p.) administration dose-dependently blocked development of morphine tolerance. DHK (10mg/kg, s.c.), administered 15 min before each morphine injection, prevented inhibition of morphine tolerance by ceftriaxone (200mg/kg, i.p.). These results identify an interaction between ceftriaxone and morphine in opioid-tolerant rats and suggest beta-lactam antibiotics preserve analgesic efficacy during chronic morphine exposure.
Sigma-Aldrich
Dihydrokainic acid, ≥98% (HPLC), powder