Merck
  • A terahertz time-domain spectrometer for simultaneous transmission and reflection measurements at normal incidence.
A terahertz time-domain spectrometer for simultaneous transmission and reflection measurements at normal incidence.
Optics express (2009-12-10)
Fabian D J Brunner, Arno Schneider, Peter Günter
ABSTRACT
We present a versatile terahertz time-domain spectrometer which allows reflection measurements at normal incidence and double pass transmission measurements in a single experimental setup. Two different modes for transmission measurements are demonstrated for precise measurements of transparent high or low refractive index materials, respectively. The refractive indices and absorption coefficients of cesium iodide, potassium bromide, sodium chloride, polytetrafluoroethylene (PTFE, Teflon), and silicon have been measured in the frequency range between 1.4 and 4.7 THz. The parameters of the Lorentz oscillator functions describing the phonon polariton dispersions of CsI and KBr have been determined.
Product Description
Supelco
Cesium iodide, analytical standard, suitable for mass spectrometry (MS)
Sigma-Aldrich
Cesium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium iodide, 99.999% trace metals basis
Sigma-Aldrich
Cesium iodide, 99.9% trace metals basis