Merck
Biosynthesis of Sb2O3 nanoparticles: a low-cost green approach.
Biotechnology journal (2009-10-22)
Anal K Jha, Kamlesh Prasad, K Prasad
ABSTRACT
A low-cost green and reproducible microbe (Lactobacillus sp.)-mediated biosynthesis of Sb(2)O(3) nanoparticles is reported. The synthesis was performed at around room temperature. X-ray and transmission electron microscopy analyses were performed to ascertain the formation of Sb(2)O(3) nanoparticles. X-ray analysis indicated that Sb(2)O(3) nanoparticles had a face-centered cubic unit cell structure. Individual nanoparticles as well as a few aggregates of 3-12 nm were found. A possible mechanism for the synthesis of nano Sb(2)O(3) is proposed.
Sigma-Aldrich
Antimony(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Antimony(III) oxide, powder, 5 μm, ReagentPlus®, 99%
Sigma-Aldrich
Antimony(III) oxide, nanopowder, <250 nm particle size (TEM), ≥99.9% trace metals basis