Merck
  • Isolation of bioactive peptides from tryptone that modulate lipase production in Yarrowia lipolytica.
Isolation of bioactive peptides from tryptone that modulate lipase production in Yarrowia lipolytica.
Bioresource technology (2009-02-04)
Saoussen Turki, Ines Ben Kraeim, Frederic Weeckers, Philippe Thonart, Héla Kallel
ABSTRACT
In this work the effect of several organic nitrogen sources on lipase production in Yarrowia lipolytica LgX64.81 overproducing mutant was studied. Among them, tryptone and peptone showed the most prominent stimulatory effect. Interestingly, only tryptic and peptic casein digest were found to highly induce lipase biosynthesis while lipase production was very limited in the presence of casein digest from papain and pronase-catalysed hydrolysis and absent in case of chymotryptic digest. It was also demonstrated that the stimulatory peptides should be present in the culture medium at specific proportions and molecular size to match the physiological requirement of Yarrowia lipolytica strain for lipase biosynthesis. Herein, the lipase-production stimulatory peptides were isolated by ion exchange chromatography for the first time. These results had contributed to gain an insight on tryptone role in lipase production by Yarrowia lipolytica. Moreover the use of a chemically defined medium supplemented with the isolated peptides, will improve the efficiency of the process for lipase production in this yeast.
Product Description
Millipore
Peptone from casein and other animal proteins, suitable for microbiology
Millipore
Peptone from milk solids, Refined hydrolysate
Millipore
Proteose Peptone, Enzymatic hydrolysate
Millipore
Peptone from animal tissue, from meat
Sigma-Aldrich
Peptone from animal tissue, from meat, BioReagent, suitable for cell culture, suitable for plant cell culture
Millipore
Peptone Primatone® RL, suitable for microbiology