Merck
  • Versatile biomimetic dendrimer templates used in the formation of TiO2 and GeO2.
Versatile biomimetic dendrimer templates used in the formation of TiO2 and GeO2.
Dalton transactions (Cambridge, England : 2003) (2008-07-17)
Sarah L Sewell, Ryan D Rutledge, David W Wright
ABSTRACT
Biomimetic synthesis is emerging as an advantageous alternative to the harsh synthetic conditions traditionally used in metal oxide syntheses techniques. Silaffins, proteins from the C. fusiformis diatom, form silica in an aqueous environment under benign conditions. Amine terminated PAMAM and PPI dendrimers are effective mimics of silaffins and other silica precipitating polyamines. We have expanded the scope of dendrimer mediated metal oxide formation to include titanium dioxide, a photocatalyst, and germanium dioxide, a blue photoluminescent material. The nanoparticles were characterized using scanning electron microscopy (SEM), Fourier transform infrared spectroscopy (IR), and X-ray diffraction patterns (XRD). A variable temperature XRD analysis of TiO(2) nanoparticles was conducted to study the transition from anatase to rutile. TiO(2) nanoparticles synthesized in phosphate buffer showed a 200 degrees C decrease in the anatase to rutile transition temperature relative to TiO(2) templated in water. XRD analysis of GeO(2) nanoparticles in either water or phosphate buffer reveal crystalline alpha-phase germanium oxide. To our knowledge, this is the first report of the synthesis of crystalline GeO(2) under ambient conditions.
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Germanium(IV) oxide, powder, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis