Merck
  • Use of Raman spectroscopy as a tool for in situ monitoring of microwave-promoted reactions.
Use of Raman spectroscopy as a tool for in situ monitoring of microwave-promoted reactions.
Nature protocols (2008-01-15)
Nicholas E Leadbeater, Jason R Schmink
ABSTRACT
The progress of microwave-promoted reactions can be monitored by interfacing a Raman spectrometer with a scientific microwave unit. The apparatus is assembled from commercially available components. It is used in this protocol to follow the base-catalyzed reaction of salicylaldehyde with ethylacetoacetate to yield 3-acetylcoumarin. It is possible to watch the reaction spectroscopically in real time, determine when it reaches completion and thus use it as a tool for rapid reaction optimization.
Sigma-Aldrich
3-Acetylcoumarin, 96%
Sigma-Aldrich
Ethyl acetoacetate, Arxada quality, ≥99.0% (GC)
Sigma-Aldrich
Ethyl acetoacetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetoacetate, natural, ≥97%, FG
Sigma-Aldrich
Ethyl acetoacetate, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl acetoacetate, puriss. p.a., ≥99.0% (GC)