Merck
  • Direct determination of chlorophenols present in liquid samples by using a supported liquid membrane coupled in-line with capillary electrophoresis equipment.
Direct determination of chlorophenols present in liquid samples by using a supported liquid membrane coupled in-line with capillary electrophoresis equipment.
Analytica chimica acta (2007-03-28)
S Almeda, L Nozal, L Arce, M Valcárcel
ABSTRACT
Actually there is a great trend on the development of effective analytical methods for monitoring trace levels of various phenols which can indicate, among others compounds, the water quality. A simple, inexpensive supported liquid membrane (SLM) device was used in combination with commercially available capillary electrophoresis (CE) equipment for the direct determination of chlorophenols in surface water samples. The manifold was used simultaneously to extract and preconcentrate the analytes from liquid samples. In the extraction set-up, the donor phase (4 mL) was placed in the CE vial, where a micro-membrane extraction unit (MMEU) accommodating the acceptor phase (100 microL) in its lumen was immersed. The supported liquid membrane was constructed by impregnating a porous Fluoropore Teflon (PTFE) membrane with a water-immiscible organic solvent (dihexyl ether). The extraction process was optimized with regard to the pH of the donor and acceptor phases, membrane liquid, extraction time and voltage applied to the inlet or outlet vial during extraction. The chlorinated phenols pentachlorophenol (PCP), 2,3,6 trichlorophenol (TCP) and 2,6 dichlorophenol (DCP) were thus efficiently separated by CE, using tris(hydroxymethyl)aminomethane (Tris) and an NaH(2)PO(4) solution containing 1% (v/v) methanol at pH 10.5 as running buffer.
Sigma-Aldrich
2,6-Dichlorophenol, 99%
Supelco
2,6-Dichlorophenol, PESTANAL®, analytical standard