Merck
  • Synthetic studies towards the phomactins. Concise syntheses of the tricyclic furanochroman and the oxygenated bicyclo[9.3.1]pentadecane ring systems in phomactin A.
Synthetic studies towards the phomactins. Concise syntheses of the tricyclic furanochroman and the oxygenated bicyclo[9.3.1]pentadecane ring systems in phomactin A.
Organic & biomolecular chemistry (2003-12-11)
Kevin M Foote, Christopher J Hayes, Matthew P John, Gerald Pattenden
ABSTRACT
A concise synthesis of the tricyclic furanochroman unit 3 found in the PAF antagonist phomactin A (1) isolated from the marine fungus Phoma sp., is described. In complementary studies, a variety of synthetic routes towards the bicyclo[9.3.1]pentadecane ring system 4 in phomactin A were explored. These studies culminated in a synthesis of the substituted ring system 79 containing all the carbon atoms and all the necessary oxygen centres for elaboration to phomactin A itself.
Product Description
Supelco
Pentadecane, analytical standard
Sigma-Aldrich
Pentadecane, ≥99%
Sigma-Aldrich
Pentadecane, ≥98.0% (GC)