Merck
  • [Raman spectroscopic study of silicon-oxygen tetrahedrons microstructure in x CaSiO3.LiBO2 solid solutions].
[Raman spectroscopic study of silicon-oxygen tetrahedrons microstructure in x CaSiO3.LiBO2 solid solutions].
Guang pu xue yu guang pu fen xi = Guang pu (2003-08-27)
Jing-lin You, Guo-chang Jiang, Qi Wen, Kuang-di Xu
ABSTRACT
Raman spectra of xCaSiO3.LiBO2 (by weight ratio, x = 0, 0.25, 0.33, 0.50) crystals were measured. The microstructure of different silicon-oxygen tetrahedrons and their abundant variations with silica contents were studied. It shows that all the silicon-oxygen tetrahedrons are isolated by Si-Ob-B bondings or free SiO4(4-). No evidence suggests that there exist considerable Si-Ob-Si bondings. The abundance of Q2, Q4 species increases with the increasing silica content, there exists barely Q3 species even at high silica content. And the sum of integrated Raman peaks for all Qi species are linearly correlated with silica concentration in crystal, which offers the possibility to analysis silica in minerals, slags, glasses and soil samples directly.
Sigma-Aldrich
Lithium metaborate, 99.995% trace metals basis
Sigma-Aldrich
Lithium metaborate, 99.9% trace metals basis
Sigma-Aldrich
Lithium metaborate, ACS reagent, ≥98.0%
Sigma-Aldrich
Lithium tetraborate, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium tetraborate, ≥99.995% trace metals basis