Merck
  • Sodium molybdate improves the phagocytic function in alloxan-induced diabetic rats.
Sodium molybdate improves the phagocytic function in alloxan-induced diabetic rats.
Chemico-biological interactions (2003-04-11)
Saraswathi Panneerselvam, Swaminathan Govindasamy
ABSTRACT
Diabetes mellitus is a disease, which virtually affects all the systems in the body including the immune system. Bacterial infections are important causes of increased morbidity and mortality in diabetic patients. In the present study, we assessed the effect of molybdate on immune responses of diabetic rats. The phagocytic potency and nitroblue tetrazolium dye reduction capacity were found to be considerably lowered whereas soluble immune complex formation enhanced significantly in diabetic rats when compared with control rats. From our findings, it is suggested that dietary intake of molybdate may have a vital role in enhancing immune functions during diabetic mellitus.
Sigma-Aldrich
Sodium molybdate dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium molybdate, ≥98%
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
Sodium molybdate, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, 99.99% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%