Merck
  • Electron and hole transfer from DNA base radicals to oxidized products of guanine in DNA.
Electron and hole transfer from DNA base radicals to oxidized products of guanine in DNA.
Radiation research (2003-02-26)
Zhongli Cai, Michael D Sevilla
ABSTRACT
An investigation of electron and hole transfer to oxidized guanine bases in DNA is reported. Guanine in DNA was preferentially oxidized by Br(2)(*-) at 298 K to 8-oxo-7,8-dihydro-guanine (8-oxo-G) and higher oxidation products. HPLC-EC analysis of irradiated DNA shows that the formation of 8-oxo-G could not be increased above the ratio of one 8-oxo-G to 127 +/- 6 bp regardless of dose. 8-oxo-G can be produced only at low levels because it is further oxidized to other species. These oxidation products of guanine have been extensively investigated and identified by others. Our electron spin resonance studies suggest that at 77 K 8-oxo-G is a trap for radiation-produced holes, but certain further oxidation products of 8-oxo-G (G(ox)) are found to be efficient electron traps. Gamma irradiation of oxidized DNA samples in frozen (D(2)O) aqueous ices and glassy 7 M LiBr solutions resulted in radicals formed by electron attachment to the G(ox) sites that were monitored by electron spin resonance spectroscopy (ESR) at 77 K. These ESR spectra suggest that those oxidation products of 8-oxo-G containing alpha-diketo groups account for the electron traps (G(ox)) in oxidized DNA with oxaluric acid a likely major trap. Electron transfer from DNA anion radicals to G(ox) was followed by monitoring of their ESR signals with time at 77 K. Using typical values for the tunneling constant beta estimates of the relative amount of G(ox) to base pairs were obtained. Radicals formed by UV photolysis of oxidized DNA in 8 M NaClO(4) glassy aqueous solutions were also investigated. The 8-oxo-G cation accounts for less than 10% of all the radicals observed after either gamma irradiation of oxidized DNA in frozen (D(2)O) aqueous solution or UV photolysis of oxidized DNA in 8 M NaClO(4) glassy aqueous solutions. We estimate hole transfer distances of about 7 +/- 1 bp at 1 min from G(*+) to 8-oxo-G.
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis