Merck
  • A fluorescent compound for glucose uptake measurements in isolated rat cardiomyocytes.
A fluorescent compound for glucose uptake measurements in isolated rat cardiomyocytes.
Canadian journal of physiology and pharmacology (2002-05-07)
S W Ball, J R Bailey, J M Stewart, C M Vogels, S A Westcott
ABSTRACT
A focus of current diabetes research is the development of insulinomimetic compounds for oral treatment of diabetes and its associated cardiac complications. Screening compounds for their potential insulinomimetic effects usually involves the use of radioactive isotopes. The focus of this study was to investigate a nonradioactive fluorescent compound for its use in screening insulinomimetic compounds. The indicator 2-(N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)amino)-2-deoxyglucose (2-NBDG) has been used by some workers to measure glucose uptake in Escherichia coli and Candida albicans. We propose that 2-NBDG will also be a suitable indicator for mammalian cell lines, in particular rat cardiomyocytes. We found that the indicator could give a reliable reproducible standard curve following appropriate dilution and is taken up by isolated cardiomyocytes. The insulinomimetic compounds vanadyl sulfate and sodium molybdate showed rates of glucose uptake similar to that of insulin. Furthermore, the rate of uptake measured for insulin using this technique (0.04 +/- 0.003 nmol x min(-1) x 10(6) cells(-1) is comparable with previous literature using 2-deoxyglucose uptake measurements on isolated myocytes (0.040 nmol x min(-1) x 10(6) cells(-1), demonstrating the validity of this fluorescent compound for glucose uptake studies.
Sigma-Aldrich
Sodium molybdate, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%, suitable for plant cell culture
Sigma-Aldrich
Sodium molybdate, ≥98%
Sigma-Aldrich
Sodium molybdate dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium molybdate dihydrate, 99.99% trace metals basis
Sigma-Aldrich
Sodium molybdate dihydrate, ≥99.5%