Merck
  • Investigation on molar mass, solubility and enzymatic fragmentation of xylans by multi-detected SEC chromatography.
Investigation on molar mass, solubility and enzymatic fragmentation of xylans by multi-detected SEC chromatography.
Bioresource technology (2001-10-17)
B Saake, T Kruse, J Puls
ABSTRACT
Four xylan samples from different origin were investigated, using a multi-detector, size exclusion, chromatographic system with two chromatographic column sets and mobile phases differing in the DMSO:water ratio. Molar mass distribution could be analysed best using a mobile phase of DMSO:water (90:10) with addition of 0.05 M LiBr, a system offering good solubilisation of the polymers and a proper chromatographic separation. SEC analysis in aqueous systems provided information on solubility and aggregation of xylans. A comparison of UV- and RI-signals in different systems gave further information on lignin impurities, which in some cases were involved in aggregation phenomena. Both analytical systems were applied to study the enzymatic fragmentation of xylans. Combining the information derived from the two systems can differentiate between the enzymatic degradation of the well-dissolved and the associated polymer fractions.
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O