Merck
  • Study of the formation equilibria of electron donor-acceptor complexes between [60]fullerene and methylbenzenes by absorption spectrometric method.
Study of the formation equilibria of electron donor-acceptor complexes between [60]fullerene and methylbenzenes by absorption spectrometric method.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2001-07-12)
S Bhattacharya, M Banerjee, A K Mukherjee
ABSTRACT
The electron donor-acceptor (EDA) interaction between [60]fullerene and three methylbenzenes, viz., durene, pentamethylbenzene and hexamethylbenzene has been studied in carbon tetrachloride medium at a number of temperatures. It has been found that these methylbenzenes form stable 1:1 EDA complexes with [60]fullerene. Charge transfer (CT) absorption bands of the complexes in the 410-460 nm region are more intense than the usual 420-700 nm absorption band of C60. The CT transition energies (hvCT) of the complexes change systematically with change in the number and position of the methyl groups in the donor molecules (methylbenzenes) and also with the donor ionisation potentials. From an analysis of this variation the electron affinity of C60 has been found to be 2.30 eV and also an inductive effect Hückel parameter of the methyl group has been determined. Formation constants (K) have been determined at three different temperatures from which the enthalpies and entropies of formation of the complexes have been determined.
Sigma-Aldrich
1,2,4,5-Tetramethylbenzene, 98%
Sigma-Aldrich
Hexamethylbenzene, 99%
Supelco
1,2,4,5-Tetramethylbenzene, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pentamethylbenzene, 98%