Merck
  • Improved procedure for the reductive acetylation of acyclic esters and a new synthesis of ethers.
Improved procedure for the reductive acetylation of acyclic esters and a new synthesis of ethers.
The Journal of organic chemistry (2000-05-18)
D J Kopecky, S D Rychnovsky
ABSTRACT
An optimized protocol for the DIBALH reductive acetylation of acyclic esters and diesters is described. This reductive acetylation procedure allows a wide variety of esters to be converted into the corresponding alpha-acetoxy ethers in good to excellent yields. It was found that, under mild acidic conditions, many alpha-acetoxy ethers can be further reduced to the corresponding ethers. This net two-step ester deoxygenation is an attractive alternative to the classical Williamson synthesis for certain ethers.
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in hexanes
Sigma-Aldrich
Diisobutylaluminum hydride solution, 25 wt. % in toluene
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in cyclohexane
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in heptane
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in toluene
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Diisobutylaluminum hydride, reagent grade