Merck
  • Differential expression of granzyme A and granzyme B proteases and their secretion by fresh rat natural killer cells (NK) and lymphokine-activated killer cells with NK phenotype (LAK-NK).
Differential expression of granzyme A and granzyme B proteases and their secretion by fresh rat natural killer cells (NK) and lymphokine-activated killer cells with NK phenotype (LAK-NK).
European journal of immunology (1992-04-01)
F Velotti, G Palmieri, D D'Ambrosio, M Piccoli, L Frati, A Santoni
ABSTRACT
Granzymes A-G are a family of serine proteases localized in the cytoplasmic granules of cytotoxic T lymphocytes (Masson, D. et al. Cell 1987. 49: 679) and granzyme A is secreted by T lymphocytes in response to antigenic stimulation. Granzyme A is also expressed by natural killer (NK) and lymphokine-activated killer (LAK) cells. Here we show that fresh rat NK cells constitutively express granzyme B and that granzyme A and granzyme B are differentially regulated in unstimulated NK cells vs. LAK cells with NK phenotype (LAK-NK cells). We also show that both granzymes A and B are secreted in a calcium-dependent manner, by NK and LAK-NK cells in response to stimuli which trigger NK cell functions.
Sigma-Aldrich
Z-Gly-Pro-Arg 7-amido-4-trifluoromethylcoumarin trifluoroacetate salt, ≥95% (HPLC), powder