Merck
  • [Effect of lipolytic and catalase activity on physico-mechanical properties of coating Polyken 980-25].
[Effect of lipolytic and catalase activity on physico-mechanical properties of coating Polyken 980-25].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2013-03-23)
Zh P Kopteva, V V Zanina, M A Boretskaia, A E Kopteva, I A Kozlova
ABSTRACT
Lipolytic and catalase activity of Pseudomonas pseudoalcaligenes 109, Rhodococcus erythropolis 102, Bacillus subtilis 138 and their association with different growth models: biofilm and plankton ones. It is shown that under biofilm conditions the fermentative activity of bacteria under study was 1.5-1.7 times higher than under plankton conditions. Monocultures of bacteria displayed much lower activity than associative ones. Changes of physico-chemical properties of the specimens of protective coating Polyken 980-25 with participation of the above bacteria have been studied. The coating breaking strength decreases by 5.9-11.8% under the effect of monocultures, and by 17.3% under the effect of association. The adhesive strength as the basic index of coating biologic resistance decreased, respectively, in mono- and associated cultures by 28.6-73.2% in respect of the control. Damaging the sticking layer of isolation coating, bacteria damage the adhesion to metal which favors its corrosion.
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein