Merck
  • Spectroscopic determination of the toxicity, absorption, reduction, and translocation of Cr(VI) in two Magnoliopsida species.
Spectroscopic determination of the toxicity, absorption, reduction, and translocation of Cr(VI) in two Magnoliopsida species.
International journal of phytoremediation (2013-03-16)
Milka O Montes, Jose R Peralta-Videa, Jason G Parsons, Baltazar Corral Diaz, Jorge L Gardea-Torresdey
ABSTRACT
Hexavalent chromium is a contaminant highly mobile in the environment that is toxic for plants at low concentrations. In this work, the physiological response of Convolvulus arvensis and Medicago truncatula plants to Cr(VI) treatments was compared. C. arvensis is a potential Cr hyperaccumulator well adapted to semiarid conditions that biotransform Cr(VI) to the less toxic Cr(III). M. truncatula is a model plant well adapted to semiarid conditions with a well studied genetic response to heavy metal stress. The results demonstrated that C. arvensis is more tolerant to Cr toxicity and has a higher Cr translocation to the leaves. The inductively coupled plasma optical emission spectroscopy results showed that C. arvensis plants treated with 10 mg Cr(VI) L(-1) accumulated 1512, 210, and 131 mg Cr kg(-1) in roots, stems, and leaves, respectively. While M. truncatula plants treated with the same Cr(VI) concentration accumulated 1081, 331, and 44 (mg Cr kg(-1)) in roots, stems, and leaves, respectively. Enzymatic assays demonstrated that Cr(VI) decreased ascorbate peroxidase activity and increased catalase activity in M. truncatula, while an opposite response was found in C. arvensis. The x-ray absorption spectroscopy studies showed that both plant species reduced Cr(VI) to the less toxic Cr(III).
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein