Merck
  • [The analysis of results of laboratory analyses considering the circannual rhythms].
[The analysis of results of laboratory analyses considering the circannual rhythms].
Klinicheskaia laboratornaia diagnostika (2013-03-14)
L V Barabash
ABSTRACT
The technique of cosinor-analysis was applied to study the oncological dynamics of hormonal and lipid status, activity of peroxide oxidation of lipids in 580 healthy males and females residing in the Western Siberia. The clear circannual rhythm of gluco-corticoid function of suprarenal glands, insular apparatus and activity of thyroid system, lipid metabolism and half-year periodicity, of activity of lipids peroxide oxidation are detected. The statistically significant differences of number of indicators in various phases of circannual rhythm are established.
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein