Merck
  • Biomonitoring study of an estuarine coastal ecosystem, the Sacca di Goro lagoon, using Ruditapes philippinarum (Mollusca: Bivalvia).
Biomonitoring study of an estuarine coastal ecosystem, the Sacca di Goro lagoon, using Ruditapes philippinarum (Mollusca: Bivalvia).
Environmental pollution (Barking, Essex : 1987) (2013-03-12)
Angela Sacchi, Catherine Mouneyrac, Claudia Bolognesi, Andrea Sciutto, Paola Roggieri, Marco Fusi, Gian Maria Beone, Ettore Capri
ABSTRACT
Coastal lagoons are constantly subjected to releases of chemical pollutants, and so organisms may be exposed to such toxicants. This study investigated through a multivariate approach the physiological status of bivalve Ruditapes philippinarum, farmed in Sacca di Goro lagoon. Biomarkers at different levels of biological organization (catalase, superoxide dismutase, genotoxicity, reburrowing behavior) were evaluated at three sites exposed to different environmental conditions. A seasonal trend was observed, and micronucleus frequency was significantly lowest at the relatively pristine reference site. Enzymatic activity toward oxyradicals be quite efficient since variations in responsiveness were not consistent. However, behavioral impairment was observed in reburrowing rates. Sediment concentrations showed low PAH levels and high natural levels of trace metals Cr and Ni. DistLM statistical analysis revealed a non-significant relationship between selected biomarkers and xenobiotics. Therefore other potentially toxic compounds in admixture at low doses may be involved in driving differing spatial distribution of physiological impairment.
Sigma-Aldrich
Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
Sigma-Aldrich
Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein