Merck
Oncolytic vaccinia virus therapy of salivary gland carcinoma.
JAMA otolaryngology-- head & neck surgery (2013-02-23)
Natalya Chernichenko, Gary Linkov, Pingdong Li, Richard L Bakst, Chun-Hao Chen, Shuangba He, Yong A Yu, Nanhai G Chen, Aladar A Szalay, Yuman Fong, Richard J Wong
ABSTRACT
To examine the therapeutic effects of an attenuated, replication-competent vaccinia virus (GLV-1h68) against a panel of 5 human salivary gland carcinoma cell lines. The susceptibility of 5 salivary gland carcinoma cell lines to infection and oncolysis by GLV-1h68 was assessed in vitro and in vivo. All 5 cell lines were susceptible to viral infection, transgene expression, and cytotoxic reactions. Three cell lines were exquisitely sensitive to infection by very low doses of GLV-1h68. Orthotopic parotid tumors exhibited more aggressive behavior compared with flank tumors. A single intratumoral injection of GLV-1h68 induced significant tumor regression without observed toxic effects in flank and parotid tumor models; controls demonstrated rapid tumor progression. These promising results demonstrate significant oncolytic activity by an attenuated vaccinia virus for infecting and lysing salivary gland carcinomas, supporting future clinical trials.
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid