Merck
  • The functional G143E variant of carboxylesterase 1 is associated with increased clopidogrel active metabolite levels and greater clopidogrel response.
The functional G143E variant of carboxylesterase 1 is associated with increased clopidogrel active metabolite levels and greater clopidogrel response.
Pharmacogenetics and genomics (2012-11-01)
Joshua P Lewis, Richard B Horenstein, Kathleen Ryan, Jeffrey R O'Connell, Quince Gibson, Braxton D Mitchell, Keith Tanner, Sumbul Chai, Kevin P Bliden, Udaya S Tantry, Cody J Peer, William D Figg, Shawn D Spencer, Michael A Pacanowski, Paul A Gurbel, Alan R Shuldiner
ABSTRACT
Carboxylesterase 1 (CES1) is the primary enzyme responsible for converting clopidogrel into biologically inactive carboxylic acid metabolites. We genotyped a functional variant in CES1, G143E, in participants of the Pharmacogenomics of Anti-Platelet Intervention (PAPI) study (n=566) and in 350 patients with coronary heart disease treated with clopidogrel, and carried out an association analysis of bioactive metabolite levels, on-clopidogrel ADP-stimulated platelet aggregation, and cardiovascular outcomes. The levels of clopidogrel active metabolite were significantly greater in CES1 143E-allele carriers (P=0.001). Consistent with these findings, individuals who carried the CES1 143E-allele showed a better clopidogrel response as measured by ADP-stimulated platelet aggregation in both participants of the PAPI study (P=0.003) and clopidogrel-treated coronary heart disease patients (P=0.03). No association was found between this single nucleotide polymorphism and baseline measures of platelet aggregation in either cohort. Taken together, these findings suggest, for the first time, that genetic variation in CES1 may be an important determinant of the efficacy of clopidogrel.
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells