Merck
  • Cytosolic retention of HtrA2 during mitochondrial protein import stress triggers the DELE1-HRI pathway.
Cytosolic retention of HtrA2 during mitochondrial protein import stress triggers the DELE1-HRI pathway.
Communications biology (2024-03-31)
Paul Y Bi, Samuel A Killackey, Linus Schweizer, Damien Arnoult, Dana J Philpott, Stephen E Girardin
ABSTRACT
Mitochondrial stress inducers such as carbonyl cyanide m-chlorophenyl hydrazone (CCCP) and oligomycin trigger the DELE1-HRI branch of the integrated stress response (ISR) pathway. Previous studies performed using epitope-tagged DELE1 showed that these stresses induced the cleavage of DELE1 to DELE1-S, which stimulates HRI. Here, we report that mitochondrial protein import stress (MPIS) is an overarching stress that triggers the DELE1-HRI pathway, and that endogenous DELE1 could be cleaved into two forms, DELE1-S and DELE1-VS, the latter accumulating only upon non-depolarizing MPIS. Surprisingly, while the mitochondrial protease OMA1 was crucial for DELE1 cleavage in HeLa cells, it was dispensable in HEK293T cells, suggesting that multiple proteases may be involved in DELE1 cleavage. In support, we identified a role for the mitochondrial protease, HtrA2, in mediating DELE1 cleavage into DELE1-VS, and showed that a Parkinson's disease (PD)-associated HtrA2 mutant displayed reduced DELE1 processing ability, suggesting a novel mechanism linking PD pathogenesis to mitochondrial stress. Our data further suggest that DELE1 is likely cleaved into DELE1-S in the cytosol, while the DELE1-VS form might be generated during halted translocation into mitochondria. Together, this study identifies MPIS as the overarching stress detected by DELE1 and identifies a novel role for HtrA2 in DELE1 processing.
Sigma-Aldrich
Anti-OMA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
o-Phenanthroline, A metal chelating agent that prevents the induction of chromosomal aberrations in streptozotocin-treated cells.