Merck
  • Tld1 is a regulator of triglyceride lipolysis that demarcates a lipid droplet subpopulation.
Tld1 is a regulator of triglyceride lipolysis that demarcates a lipid droplet subpopulation.
The Journal of cell biology (2023-10-27)
Natalie Ortiz Speer, R Jay Braun, Emma Grace Reynolds, Alicja Brudnicka, Jessica M J Swanson, W Mike Henne
ABSTRACT
Cells store lipids in the form of triglyceride (TG) and sterol ester (SE) in lipid droplets (LDs). Distinct pools of LDs exist, but a pervasive question is how proteins localize to and convey functions to LD subsets. Here, we show that the yeast protein YDR275W/Tld1 (for TG-associated LD protein 1) localizes to a subset of TG-containing LDs and reveal it negatively regulates lipolysis. Mechanistically, Tld1 LD targeting requires TG, and it is mediated by two distinct hydrophobic regions (HRs). Molecular dynamics simulations reveal that Tld1's HRs interact with TG on LDs and adopt specific conformations on TG-rich LDs versus SE-rich LDs in yeast and human cells. Tld1-deficient yeast display no defect in LD biogenesis but exhibit elevated TG lipolysis dependent on lipase Tgl3. Remarkably, overexpression of Tld1, but not LD protein Pln1/Pet10, promotes TG accumulation without altering SE pools. Finally, we find that Tld1-deficient cells display altered LD mobilization during extended yeast starvation. We propose that Tld1 senses TG-rich LDs and regulates lipolysis on LD subpopulations.
Sigma-Aldrich
Monoclonal Anti-HSP90B1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2647, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture